Recombinant Mouse CCL2 (from E. coli), His-tag
Catalog # ABNOP9510.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:P10148
- Protein/peptide name:CCL2
- Sequence:24a.a.-148a.a.