Order Entry
Export
ContactUsLinkComponent
Recombinant Human GMFB (from E. coli)
Recombinant Human GMFB (from E. coli)
Catalog # ABNOP8750.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human GMFB (from E. coli)
Catalog # ABNOP8750.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    GMFB

Specifications

About this item