Order Entry
Export
ContactUsLinkComponent
Recombinant Human IL22 (from E. coli)
Recombinant Human IL22 (from E. coli)
  ABNOP8801.10UG
 :  Abnova
 :  
Recombinant Human IL22 (from E. coli)
  ABNOP8801.10UG
 :  Abnova
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    Q9GZX6
  • Protein/peptide name:
    IL22