Order Entry
Export
ContactUsLinkComponent
Recombinant Human EBI3 (from E. coli)
Recombinant Human EBI3 (from E. coli)
Catalog # ABNOP8541.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Human EBI3 (from E. coli)
Catalog # ABNOP8541.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    Q14213
  • Protein/peptide name:
    EBI3

Specifications

About this item