Order Entry
Export
ContactUsLinkComponent
Recombinant Human EREG (from E. coli)
Recombinant Human EREG (from E. coli)
  ABNOP8568.25UG
 :  Abnova
 :  
Recombinant Human EREG (from E. coli)
  ABNOP8568.25UG
 :  Abnova
 :  

 

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    O14944
  • Protein/peptide name:
    EREG