Order Entry
Export
ContactUsLinkComponent
Recombinant Human CD48 (from Baculovirus), His-tag
Recombinant Human CD48 (from Baculovirus), His-tag
Catalog # ABNOP8024.100UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CD48 (from Baculovirus), His-tag
Catalog # ABNOP8024.100UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    P09326
  • Protein/peptide name:
    CD48
  • Sequence:
    27a.a.-220a.a.

Specifications

About this item