Order Entry
Export
ContactUsLinkComponent
Recombinant Human CTSZ (from Baculovirus), His-tag
Recombinant Human CTSZ (from Baculovirus), His-tag
  ABNOP8055.100UG
 :  Abnova
 :  
Recombinant Human CTSZ (from Baculovirus), His-tag
  ABNOP8055.100UG
 :  Abnova
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    Q9UBR2
  • Protein/peptide name:
    CTSZ
  • Sequence:
    24a.a.-303a.a.