Order Entry
Export
ContactUsLinkComponent
Recombinant Human VTN (from Baculovirus), His-tag
Recombinant Human VTN (from Baculovirus), His-tag
Catalog # ABNOP8056.100UG
Supplier:  Abnova
CAS Number:  
Recombinant Human VTN (from Baculovirus), His-tag
Catalog # ABNOP8056.100UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    P04004
  • Protein/peptide name:
    VTN
  • Sequence:
    20a.a.-478a.a.

Specifications

About this item