Order Entry
Export
ContactUsLinkComponent
Recombinant Human ERGIC3 (from E. coli), His-tag
Recombinant Human ERGIC3 (from E. coli), His-tag
Catalog # ABNOP7780.500UG
Supplier:  Abnova
CAS Number:  
Recombinant Human ERGIC3 (from E. coli), His-tag
Catalog # ABNOP7780.500UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_057050
  • Protein/peptide name:
    ERGIC3
  • Sequence:
    47a.a.-341a.a

Specifications

About this item