Order Entry
Export
Orders LinkContactUsLinkComponent
Recombinant Human CCL3 (from E. coli)

Recombinant Human CCL3 (from E. coli)

1 mg
Catalog # ABNOP7623.1MG
Supplier:   Abnova
Recombinant Human CCL3 (from E. coli)
Catalog # ABNOP7623.1MG
Supplier:  Abnova
CAS Number:  
Recombinant Human CCL3 (from E. coli)

1 mg

CAS Number:  
Price
4 540,00 €
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    ABNOP7623.1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    CCL3
  • Gene ID:
    P10147
  • Sequence:
    24a.a.-92a.a.
  • Species:
    Human
  • Source:
    E. coli

Specifications