Recombinant Human CXCL1 (from E. coli)
Catalog # ABNOP7498.5UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P09341
- Protein/peptide name:CXCL1
- Sequence:35a.a.-107a.a.