0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Proteins and Peptides
Recombinant Human IL15 (from E. coli)
Recombinant Human IL15 (from E. coli)
ABNOP7502.10UG
:
Abnova
:
Recombinant Human IL15 (from E. coli)
ABNOP7502.10UG
:
Abnova
:
Pk:
10 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Gene ID:
P40933
Protein/peptide name:
IL15
Sequence:
49a.a.-162a.a.