Recombinant Human ACE2(HUMAN) (from CHO cells)
Catalog # ABNOP7399.1MG
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Gene ID:Q9BYF1
- Protein/peptide name:ACE2(HUMAN)
- Sequence:18a.a.-740a.a.