Order Entry
Export
ContactUsLinkComponent
 
Recombinant Human CCL5 (from HEK293 cells)
  ABNOP7393.5UG
 :  Abnova
 :  
Recombinant Human CCL5 (from HEK293 cells)
  ABNOP7393.5UG
 :  Abnova
 :  

 

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Gene ID:
    P13501
  • Protein/peptide name:
    CCL5
  • Sequence:
    24a.a.-91a.a.