Recombinant Human IL4R (from HEK293 cells)
Catalog # ABNOP7408.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Gene ID:P24394
- Protein/peptide name:IL4R
- Sequence:24a.a.-232a.a.