0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Proteins and Peptides
Recombinant Rat LEP (from E. coli)
Recombinant Rat LEP (from E. coli)
Catalog # ABNOP7414.5MG
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Rat LEP (from E. coli)
Catalog # ABNOP7414.5MG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
5 mg
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Rat
Gene ID:
P50596
Protein/peptide name:
LEP
Sequence:
22a.a.-167a.a.
Specifications
About this item
Product Details
Product Details