Recombinant Mouse CSF3 (from CHO cells)
Catalog # ABNOP7417.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Gene ID:P09920
- Protein/peptide name:CSF3
- Sequence:31a.a.-208a.a.