Order Entry
Export
Orders LinkContactUsLinkComponent
Recombinant Human CSF2 (from CHO cells)

Recombinant Human CSF2 (from CHO cells)

10 µG
Catalog # ABNOP7110.10UG
Supplier:   Abnova
Recombinant Human CSF2 (from CHO cells)
Catalog # ABNOP7110.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CSF2 (from CHO cells)

10 µG

CAS Number:  
Price
226,00 €
Log in for availability
1 * 10 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    ABNOP7110.10UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    CSF2
  • Gene ID:
    P04141
  • Sequence:
    a.
  • Species:
    Human
  • Source:
    CHO cells

Specifications