Anti-TNFRSF9 Rabbit Recombinant Antibody [clone: LOB12.3]
Catalog # ABNORAB03647.200UG
Supplier: Abnova
Specifications
- Antigen symbol:TNFRSF9
- Clonality:Recombinant
- Clone:LOB12.3
- Host:Rabbit
- Reactivity:Mouse
- Immunogen:Recombinant rabbit monoclonal antibody raised against mouse Tnfrsf9.
- Pk:200 µG