Anti-NCR3LG1 Rabbit Recombinant Antibody [clone: 17B1.3]
ABNORAB03669.200UG
: Abnova
- Antigen symbol:NCR3LG1
- Clonality:Recombinant
- Clone:17B1.3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human NCR3LG1.
- Pk:200 µG