Order Entry
Export
ContactUsLinkComponent
 
Anti-ADORA2A Mouse Recombinant Antibody [clone: IG2838]
Catalog # ABNORAB03653.200UG
Supplier:  Abnova
Anti-ADORA2A Mouse Recombinant Antibody [clone: IG2838]
Catalog # ABNORAB03653.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    ADORA2A
  • Clonality:
    Recombinant
  • Clone:
    IG2838
  • Host:
    Mouse
  • Reactivity:
    Human
  • Immunogen:
    Recombinant mouse monoclonal antibody raised against human ADORA2A.
  • Pk:
    200 µG

Specifications

About this item