Order Entry
Export
ContactUsLinkComponent
Anti-E Cadherin Rabbit Recombinant Antibody
Anti-E Cadherin Rabbit Recombinant Antibody
Catalog # ABNORAB02404.100UL
Supplier:  Abnova
Anti-E Cadherin Rabbit Recombinant Antibody
Catalog # ABNORAB02404.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    E Cadherin
  • Clonality:
    Recombinant
  • Host:
    Rabbit
  • Immunogen:
    Rabbit monoclonal antibody raised against recombinant E Cadherin.
  • Pk:
    100 µl

Specifications

About this item