Anti-MAPK14 Rabbit Recombinant Antibody [clone: P38T180Y182-E3]
Catalog # ABNORAB02780.200UL
Supplier: Abnova
Specifications
- Antigen symbol:MAPK14
- Clonality:Recombinant
- Clone:P38T180Y182-E3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human MAPK14.
- Pk:200 µl