Anti-EIF4EBP1 Rabbit Recombinant Antibody [clone: 4EB1T37T46-A5]
Catalog # ABNORAB02786.200UL
Supplier: Abnova
Specifications
- Antigen symbol:EIF4EBP1
- Clonality:Recombinant
- Clone:4EB1T37T46-A5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human EIF4EBP1.
- Pk:200 µl