Anti-RB1 Rabbit Recombinant Antibody [clone: RBS807S811-D9]
Catalog # ABNORAB02788.200UL
Supplier: Abnova
Specifications
- Antigen symbol:RB1
- Clonality:Recombinant
- Clone:RBS807S811-D9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human RB1.
- Pk:200 µl