Order Entry
Export
ContactUsLinkComponent
Anti-SERPINE1 Rabbit Recombinant Antibody [clone: R05-6J6]
Anti-SERPINE1 Rabbit Recombinant Antibody [clone: R05-6J6]
Catalog # ABNORAB02026.100UL
Supplier:  Abnova
Anti-SERPINE1 Rabbit Recombinant Antibody [clone: R05-6J6]
Catalog # ABNORAB02026.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    SERPINE1
  • Clonality:
    Recombinant
  • Clone:
    R05-6J6
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human SERPINE1.
  • Pk:
    100 µl

Specifications

About this item