Anti-TP53 Rabbit Recombinant Antibody [clone: R07-6A7]
Catalog # ABNORAB02031.100UL
Supplier: Abnova
Specifications
- Antigen symbol:TP53
- Clonality:Recombinant
- Clone:R07-6A7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TP53.
- Pk:100 µl