Anti-PTGES Rabbit Recombinant Antibody [clone: R03-8E7]
ABNORAB01981.100UL
: Abnova
- Antigen symbol:PTGES
- Clonality:Recombinant
- Clone:R03-8E7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human PTGES.
- Pk:100 µl