Anti-PRKACG Rabbit Recombinant Antibody [clone: R01-8I1]
ABNORAB01984.100UL
: Abnova
- Antigen symbol:PRKACG
- Clonality:Recombinant
- Clone:R01-8I1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human PRKACG.
- Pk:100 µl