Order Entry
Export
ContactUsLinkComponent
Anti-ALPP Rabbit Recombinant Antibody [clone: R01-8I3]
Anti-ALPP Rabbit Recombinant Antibody [clone: R01-8I3]
Catalog # ABNORAB01991.100UL
Supplier:  Abnova
Anti-ALPP Rabbit Recombinant Antibody [clone: R01-8I3]
Catalog # ABNORAB01991.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    ALPP
  • Clonality:
    Recombinant
  • Clone:
    R01-8I3
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human ALPP.
  • Pk:
    100 µl

Specifications

About this item