Order Entry
Export
ContactUsLinkComponent
Anti-COL3A1 Rabbit Recombinant Antibody [clone: R06-4E1]
Anti-COL3A1 Rabbit Recombinant Antibody [clone: R06-4E1]
Catalog # ABNORAB01283.100UL
Supplier:  Abnova
Anti-COL3A1 Rabbit Recombinant Antibody [clone: R06-4E1]
Catalog # ABNORAB01283.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    COL3A1
  • Clonality:
    Recombinant
  • Clone:
    R06-4E1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human COL3A1.
  • Pk:
    100 µl

Specifications

About this item