Order Entry
Export
ContactUsLinkComponent
Anti-GSDME Rabbit Recombinant Antibody [clone: R07-5I1]
Anti-GSDME Rabbit Recombinant Antibody [clone: R07-5I1]
  ABNORAB01260.100UL
 :  Abnova
Anti-GSDME Rabbit Recombinant Antibody [clone: R07-5I1]
  ABNORAB01260.100UL
 :  Abnova

 

  • Antigen symbol:
    GSDME
  • Clonality:
    Recombinant
  • Clone:
    R07-5I1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human GSDME.
  • Pk:
    100 µl