Order Entry
Export
ContactUsLinkComponent
Anti-FTO Rabbit Recombinant Antibody [clone: R06-8G5]
Anti-FTO Rabbit Recombinant Antibody [clone: R06-8G5]
Catalog # ABNORAB01265.100UL
Supplier:  Abnova
Anti-FTO Rabbit Recombinant Antibody [clone: R06-8G5]
Catalog # ABNORAB01265.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    FTO
  • Clonality:
    Recombinant
  • Clone:
    R06-8G5
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human FTO.
  • Pk:
    100 µl

Specifications

About this item