Order Entry
Export
ContactUsLinkComponent
Anti-TRIM21 Rabbit Recombinant Antibody [clone: R01-1E8]
Anti-TRIM21 Rabbit Recombinant Antibody [clone: R01-1E8]
Catalog # ABNORAB01201.100UL
Supplier:  Abnova
Anti-TRIM21 Rabbit Recombinant Antibody [clone: R01-1E8]
Catalog # ABNORAB01201.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    TRIM21
  • Clonality:
    Recombinant
  • Clone:
    R01-1E8
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human TRIM21.
  • Pk:
    100 µl

Specifications

About this item