Anti-CAV1 (aa 1-104) Mouse Monoclonal Antibody [clone: (PAT4C1AT)]
ABNOMAB23659.20UG
: Abnova
- Antigen symbol:CAV1 (aa 1-104)
- Clonality:Monoclonal
- Clone:(PAT4C1AT)
- Host:Mouse
- Reactivity:Human
- Immunogen:Mouse monoclonal antibody raised against human CAV1 amino acids 1-104 purified from E. coli.
- Pk:20 µG