Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-KLH Mouse Monoclonal Antibody [clone: (PMYCSHG)]
Anti-KLH Mouse Monoclonal Antibody [clone: (PMYCSHG)]
Anti-KLH Mouse Monoclonal Antibody [clone: (PMYCSHG)]
ABNOMAB23661.100UG
:
Abnova
Anti-KLH Mouse Monoclonal Antibody [clone: (PMYCSHG)]
ABNOMAB23661.100UG
:
Abnova
Antigen symbol:
KLH
Clonality:
Monoclonal
Clone:
(PMYCSHG)
Host:
Mouse
Immunogen:
Mouse monoclonal antibody raised against KLH.
Pk:
100 µG