Sentinel XE® LiPO Battery Charging Kit
Catalog # ILCDS-4504EU
Supplier: ILC DOVER
Specifications
- Description:LiPO battery charging kit, EU plug adapter
- Seq:01
- Pk:1
Specifications
About this item
Charges Sentinel XE® LiPO rechargeable battery.
- Easy to use charger
- Battery slides in and securely engages
- Available with EU or UK plug adapter
Charging takes approximately 5 hours if battery is completely drained. Contains cradle and plug adapter.