ContactUsLinkComponent
Sentinel® Extendible Breathing Tube
Sentinel® Extendible Breathing Tube
Catalog # ILCDS-3506
Supplier:  ILC DOVER
undefined
Sentinel® Extendible Breathing Tube
Catalog # ILCDS-3506
Supplier:  ILC DOVER
Specifications
  • Description:
    Extendible breathing tube
  • Pk:
    1
Specifications
About this item
With minimum length of 48 cm, this flexible breathing tube can be extended to 72 cm to accommodate a wide range of heights and movements.
  • Flexible breathing tube
  • Easily extends to support a wide range of motions