0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Specific Gravity Bottles
Accessories for Pycnometers
Accessories for Pycnometers
Bottle for pycnometer NS60/46, 1000 ml
LENZ07690070
:
Lenz Laborglas
Accessories for Pycnometers
LENZ07690070
:
Lenz Laborglas
Accessories for Pycnometers
Bottle for pycnometer NS60/46, 1000 ml
62,40 €
1 * 1 items
-
+
Description:
Bottle for pycnometer NS60/46, 1000 ml