Orders LinkContactUsLinkComponent
TIG Welding Gauntlets
TIG Welding Gauntlets
Supplier:   Portwest
TIG Welding Gauntlets
Supplier:  Portwest
Certificates
TIG Welding Gauntlets
3 Options Available Below
Product Details & Documents
The TIG Ultra Welding Gauntlet offers fantastic dexterity with a goat leather palm and excellent burn resistance due to the split leather backing. Designed for the professional TIG welder with aramid stitching for ultra durability.
  • EN12477 Type A welding protection
  • Premium quality leather welding gauntlet
  • The ultimate in dexterity for welders
  • Made from high quality durable goat leather
Ideal for welding and metal handling, these gloves feature reinforced Aramid stitching for maximum durability and burn protection.
Colour: Brown
 : EN ISO 21420: 2020 Dexterity 2, EN 388: 2016 + A1: 2018 3243X, EN 12477: 2001 Type A, EN 407: 2020 422X31. ANSI/ISEA 105: 2016 CUT Level A2. CE certified.
Specifications for Product Family
Specifications
  • Catalog No:
  • PRTWA521BRRL
  • PRTWA521BRRXL
  • PRTWA521BRRXXL
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Leather/polyester
  • Leather/polyester
  • Leather/polyester
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Availability
    Price
  • L
    Brown
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 15,80 €
    15,80 €
    1 * 1 PAIR
       15,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWA521BRRL
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Leather/polyester
    Sterility
    Non sterile
  • XL
    Brown
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 15,80 €
    15,80 €
    1 * 1 PAIR
       15,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWA521BRRXL
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Leather/polyester
    Sterility
    Non sterile
  • XXL
    Brown
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 15,80 €
    15,80 €
    1 * 1 PAIR
       15,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWA521BRRXXL
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Leather/polyester
    Sterility
    Non sterile