ContactUsLinkComponent
Disconnectors, Tapered
Catalog # KART7053700
Supplier:  KARTELL
undefined
Disconnectors, Tapered
Catalog # KART7053700
Supplier:  KARTELL
Specifications
  • Ext.Ø:
    14-15-16 mm
  • Description:
    Disconnectors
  • Pk:
    100
Specifications
About this item
These quick disconnector is useful for joining hoses of glass, rubber or plastic. Connectors fit tightly together. Replace expensive adapters.
Dimensions are shown as both nominal and exact as elasticity of hoses will influence the choice of size.
Accessories information: Tested according to reach regulation 1907/2006 Annex XVII.