ContactUsLinkComponent
Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™
Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™
Catalog # OXOIR4716000
CAS Number:  
undefined
Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™
Catalog # OXOIR4716000
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™
Specifications
About this item
Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.