Culti-Loops™ Aspergillus fumigatus ATCC™ 204305™
Catalog # OXOIR4601311
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:5
- Description:Aspergillus fumigatus ATCC™ 204305™
- Seq:58
Specifications
About this item
Culti-Loops™ QC organisms. Aspergillus fumigatus ATCC™ 204305™ is recommended to be used for antifungal susceptibility testing quality control. The strain is recommended for antifungal MIC applications QC testing by EUCAST.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.