ContactUsLinkComponent
Culti-Loops™ Proteus mirabilis 511-116
Culti-Loops™ Proteus mirabilis 511-116
Catalog # OXOIR4608088
CAS Number:  
undefined
Culti-Loops™ Proteus mirabilis 511-116
Catalog # OXOIR4608088
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Proteus mirabilis 511-116
Specifications
About this item
Confidently carry out testing of media, reagents, and identification and antimicrobial susceptibility kits and evaluate the bacteriological procedures with Culti-Loops™ ready-to-use QC organisms. Proteus mirabilis 511-116 strain produces extended-spectrum β-lactamase (ESBL). This strain is recommended to be used as a positive control strain with Spectra™ ESBL Medium following the HAI surveillance programs.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Caution: For in vitro diagnostic use.