Culti-Loops™ Escherichia coli ATCC™ BAA-201™
Catalog # OXOIR4609482
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:5
- Description:Escherichia coli ATCC™ BAA-201™
Specifications
About this item
Confidently carry out testing of media, reagents, and identification and antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Culti-Loops™ QC organisms.
The loop can be streaked directly onto appropriate media. Alternatively, it may be dissolved in a liquid medium.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.