3,5-Difluoro-2-hydroxyphenylboronic acid
Catalog # APOSPC412053-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:173.91 g/mol
CAS Number:1150114-51-4
:MFCD17011981
Formula:C₆H₅BF₂O₃
MDL Number:MFCD03095356