Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluoro-4-hydroxyphenylboronic acid
3,5-Difluoro-4-hydroxyphenylboronic acid
3,5-Difluoro-4-hydroxyphenylboronic acid
3,5-Difluoro-4-hydroxyphenylboronic acid
Supplier:
Apollo Scientific
Certificates
Expand image
3,5-Difluoro-4-hydroxyphenylboronic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
1132666-81-9
:
MFCD17011981
Formula:
C₆H₅BF₂O₃
:
173.91 g/mol