Soybean oil USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:8001-22-7
:
MDL Number:MFCD00132356
Density:0,917
Storage Temperature:Ambient
Specification Test Results
- Alkaline impurities:Passes test
- Acid value:Max. 0.3
- Peroxide value:Max. 10.0
- Fatty acid composition:Passes test
- Heavy metals:Max. 10 µg/g
- Sterol composition (as Brassicasterol):Max. 0.3 %
- Water:Max. 0.1%
- Unsaponifiable matter:Max.1.5%
- Identification:Passes test
- Residual solvents:Passes test