Acid Red 18 (Food Red No. 102)
Catalog # SPCMCO102-50KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Fiber drum
- Pk:50 kg
- Seq:4418
Specifications
Specification Test Results
- Absorption Max:506 (335) NM
Specification Test Results
About this item
:
:604.46 g/mol
Storage Temperature:Ambient
CAS Number:2611-82-7
Formula:C₂₀H₁₄N₂O₁₀S₃
MDL Number:MFCD00004084