Triacetin (glycerol triacetate) ≥97% USP
Catalog # SPCMTR106-500MLGL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Assay:97% Min.
Specification Test Results
About this item
:
:218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:4 °C
Density:1,161
Flash Pt:138 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-051-9
CAS Number:102-76-1
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716